missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ FL L-Cystine

Product Code. 10193712
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10193712 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10193712 Supplier Invitrogen™ Supplier No. B20340
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

BODIPY™ FL L-cystine

We have attached two BODIPY FL fluorophores to the amino groups of the disulfide-containing amino acid cystine to create this reagent for reversible thiol-specific labeling of thiolated oligonucleotides, proteins and cells. BODIPY FL L-cystine is virtually nonfluorescent due to interactions between the two fluorophores; however, thiol-specific exchange with thiolated biomolecules occurs to form mixed disulfides, resulting in green fluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type FL L-Cystine
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Thiol
Label or Dye BODIPY™ FL
Label Type BODIPY Dyes
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.