missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BODIPY™ TR Cadaverine (5-(((4-(4,4-Difluoro-5-(2-Thienyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-yl)phenoxy)acetyl)amino)pentylamine, Hydrochloride)
Description
The amine-containing BODIPY™ TR cadaverine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247). The electronically neutral BODIPY™ TR dye is spectrally similar to the red fluorescent Texas Red™™ dye.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | TR Cadaverine |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Carboxylic Acid, Ketone, Aldehyde |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Amine, Cadaverine |
| Label or Dye | BODIPY™ TR |
| Label Type | BODIPY Dyes |
| Product Line | BODIPY |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?