missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ BOLA™ Screw Caps with Aperture, PBTP
Click to view available options
Thread Size (Metric):
GL 25
Unit Size:
Pack of 5
Description
- Screw cap with handy knurl
- Suitable for GL threads
- With aperture
- Made of PBTP
Specifications
Specifications
| Diameter (English) | 0.6 in. |
| Material | PBT |
| Thread Size (Metric) | GL 25 |
| Neck Finish | 98 mm |
| Diameter (Metric) | 15 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction