missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Spare Washer for Stopcocks

Product Code. 30274468
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30274468 Polypropylene Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30274468 Supplier Bohlender™ Supplier No. J119821
  • ONLINE EXCLUSIVE PRICE
    53.48 €
    41.00 € / Pack of 50
    Save 12.48 €
    23% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spare washer for stopcocks made of polypropylene or PTFE.

  • Washers made of PP or PTFE
  • For BOLA™ Eco Stopcock Plugs
  • Color: Red

Specifications

Type Thread M 8
Description Taper 1:10/1:10/1:5
Material Polypropylene
Closure Size NS 12.5, NS 14.5, 13.5
For Use With (Equipment) For BOLA™ Eco Stopcock Plugs
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.