Learn More
Emma Safety Footwear Bologna Brogue-Style Business Shoes
With ankle support in the shank.
Brand:  Emma Safety Footwear MM10709043
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10526944
  • 168.00 € / 1 pair
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Polished black full-grain leather uppers
  • Impenetrable nitrile rubber sole
  • Hydro-Tec™ sanitized silver and wellness leather inlay sole
  • Woven zero penetration insole
  • Anti-bacterial with leather top layer in-lay sole
  • Antistatic properties: ESD 0,1 - 35 M Ohm / NEN-EN-IEC 61340-5-1
Specifications
Specifications
Black
Ankle support
Anti-bacterial with leather top layer inlay sole
Brogue
Business Shoe
Leather
43
Impenetrable nitrile rubber
EN ISO 20345:2011; ESD 0,1-35 M Ohm/NEN-EN-IEC 61340-5-1