missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Empty Mini Gel Cassette Combs, 10-well

Product Code. 15304614
Change view
Click to view available options
Quantity:
20 combs
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15304614 20 combs Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15304614 Supplier Invitrogen™ Supplier No. NW3010
  • ONLINE EXCLUSIVE PRICE
    27.53 €
    22.30 € / Pack of 20
    Save 5.23 €
    18% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used with Bolt™ Empty Minigel Cassettes (Mfr. No. NW2010) for use in gel electrophoresis. Bolt™ Minigel Cassette Combs give you the option of pouring your own special gel type while still enjoying the advantages of the Mini Gel Tank.

Bolt Empty Gel Cassette Combs are for use with Bolt Empty Gel Cassettes, which give you the option of pouring your own special gel type while still enjoying the advantages of the Mini Gel Tank and XCell SureLock Mini-Cell. All cassettes are sealed on three sides (so no gel leakage) and feature the unique wedge-shaped well design. The cassettes and combs are available in the standard 1-mm thickness. The combs are available for 10 or 12 wells.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mini Gel Tank and XCell SureLock Mini-Cell
Gel Size Mini (8 x 8 cm)
Product Line Bolt
Type Empty Mini Gel Cassette Comb
Quantity 20 combs
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.