missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Empty Mini Gel Cassettes

Product Code. 15394604
Change view
Click to view available options
Quantity:
20 cassettes
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15394604 20 cassettes Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15394604 Supplier Invitrogen™ Supplier No. NW2010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows users to pour custom gel formulations while still enjoying the advantages of the Mini Gel Tank

Bolt Empty Gel Cassettes give you the option of pouring your own special gel type while still enjoying the advantages of the Mini Gel Tank and XCell SureLock Mini-Cell. The cassettes are sealed on three sides (so no gel leakage) and feature the unique wedge-shaped well. They are available in the standard 1-mm thickness and can be used with Bolt™ 10- or 12-well combs (Cat. No. NW3010 and NW3012). Since Bolt™ empty cassettes are pre-sealed with the slot pre-taped, a bulky casting stand is not necessary.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mini Gel Tank and XCell SureLock Mini-Cell
Gel Size Mini (8 x 8 cm)
Product Line Bolt
Type Empty Mini Gel Cassette
Quantity 20 cassettes
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.