missing translation for 'onlineSavingsMsg'
Learn More

Berkshire International™ Bond 104 Cellulose Papers
BIOPHARMA

Product Code. 13485409 Shop All Berkshire Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
13485409 White Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13485409 Supplier Berkshire International™ Supplier No. B104A4WH4HP2501
  • ONLINE EXCLUSIVE PRICE
    581.85 €
    392.96 € / Pack of 10
    Save 188.89 €
    32% Off
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Material Cellulose
Color White
For Use With (Application) Writing, Copying, and Printing Within Controlled Environments
For Use With (Equipment) Photocopiers and Laser Printers
Page Format Sheets
Product Type Paper
Size A4
Weight (Metric) 104 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.