missing translation for 'onlineSavingsMsg'
Learn More

Berkshire International™ BCR™ Bond Paper
BIOPHARMA

Product Code. 12837143 Shop All Berkshire Products
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Size:
A3
A4
A5
Product Type:
BCR Bond 1040
BCR Bond 680
BCR Bond 850
Unit Size:
Pack of 250
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Size Product Type Unit Size
12837143 Green A4 BCR Bond 850 Pack of 250
12847143 White A4 BCR Bond 680 Pack of 250
12857143 Blue A4 BCR Bond 680 Pack of 250
12897133 White A4 BCR Bond 850 Pack of 250
12807143 Blue A4 BCR Bond 850 Pack of 250
12827143 Yellow A4 BCR Bond 850 Pack of 250
12817143 Pink A4 BCR Bond 850 Pack of 250
12877133 White A3 BCR Bond 850 Pack of 250
12887133 Blue A3 BCR Bond 850 Pack of 250
12867133 White A5 BCR Bond 1040 Pack of 250
12827133 White A4 BCR Bond 1040 Pack of 250
12837133 Blue A4 BCR Bond 1040 Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 12837143 Leverandør Berkshire International™ Leverandørnr. B85A4GN25010
  • ONLINE EXCLUSIVE PRICE
    29.72 €
    21.78 € / Pack of 250
    Save 7.94 €
    26% Off
In Stock
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Our BCR™ Bond is made with the purest quality cellulose from specially selected forests and is specially coated to minimize particle and fiber release.

  • Available in 5 colors: Green, Blue, Pink, White, and Yellow

cleanroom, paper, bond

Specifications

Material Cellulose
Color Green
For Use With (Application) Writing, Copying, and Printing Within Controlled Environments
For Use With (Equipment) Photocopiers and Laser Printers
Page Format Sheets
Product Type BCR Bond 850
Size A4
Weight (Metric) 850 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.