missing translation for 'onlineSavingsMsg'
Learn More

Syngene Bonded Carbon Filter
GREENER_CHOICE

Product Code. 17745924 Shop All Syngene Products
Change view
Click to view available options
Model:
ACF121
ACF12118
ACF141
ACF14118
ACF1G1
ACF1G118
ACF43F
ACFGFG
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
17745924 ACF121 Each
17755924 ACF12118 Each
17795924 ACF141 Each
17705934 ACF14118 Each
17755934 ACF1G1 Each
17765934 ACF1G118 Each
17785964 ACF43F Each
17775984 ACFGFG Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17745924 Supplier Syngene Supplier No. SYNACF121

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AirClean™ Systems bonded carbon filters provide for a predictable and reliable gas phase filtration solution.

  • Bonded layered carbon filter for organics and inorganics
  • Even distribution of impregnates, if applicable
  • Multi-layering options for larger application compatibility

Specifications

Filter Type Carbon
For Use With (Equipment) Airclean
Product Type Carbon Filter
Material Activated Carbon
Model ACF121
For Use With (Application) Chemical extraction
Color Silver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.