missing translation for 'onlineSavingsMsg'
Learn More

Borane-dimethyl sulfide complex, 94%

Product Code. 11446750
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446750 25 mL 25mL
11456750 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446750 Supplier Thermo Scientific Alfa Aesar Supplier No. L07705.AC
 Restricted Item
In Stock
Add to basket
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

CAS: 13292-87-0 | C2H9BS | 75.96 g/mol

Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.

Solubility
Miscible with ethyl ether, dichloromethane, benzene, xylene, hexane, diglyme, dimethyl ether, ethyl acetate, toluene, methylene chloride and other aprotic solvents.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Store in a cool place. Incompatible with acids, water, alcohols, bromine, acid anhydrides, acid chlorides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 13292-87-0
Molecular Formula C2H9BS
Molecular Weight (g/mol) 75.96
MDL Number MFCD00013189
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
PubChem CID 9833925
IUPAC Name boron;methylsulfanylmethane
SMILES B.CSC

Spezifikation

Melting Point -38°C
Density 0.801
Boiling Point ∼44°C (decomposition)
Flash Point −1°C (30°F)
Linear Formula (CH3)2S·BH3
Refractive Index 1.457
Quantity 25 mL
UN Number UN2924
Beilstein 3663489
Sensitivity Air and moisture sensitive
Solubility Information Miscible with ethyl ether,dichloromethane,benzene,xylene,hexane,diglyme,dimethyl ether,ethyl acetate,toluene,methylene chloride and other aprotic solvents.
Formula Weight 75.97
Percent Purity 94%
Chemical Name or Material Borane-dimethyl sulfide complex
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.