missing translation for 'onlineSavingsMsg'
Learn More

Borane-tert-butylamine complex, 97%

Product Code. 11367956
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367956 10 g 10g
11377956 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367956 Supplier Thermo Scientific Alfa Aesar Supplier No. L13177.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7337-45-3 | C4H14BN | 86.97 g/mol

Borane-tert-butylamine complex is used in stereoselective reduction of a steroidal ketone and for a review of amine boranes as selective reducing and hydroborating agents. It is also used as strong co-reducing agent in the syntheses of copper (I) oxide nanoparticles, diphosphine-protected gold nanocluster and 4-acetoxycinnamyl alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borane-tert-butylamine complex is used in stereoselective reduction of a steroidal ketone and for a review of amine boranes as selective reducing and hydroborating agents. It is also used as strong co-reducing agent in the syntheses of copper (I) oxide nanoparticles, diphosphine-protected gold nanocluster and 4-acetoxycinnamyl alcohols.

Solubility
Soluble in water (27 g/L at 20°C) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7337-45-3
Molecular Formula C4H14BN
Molecular Weight (g/mol) 86.97
MDL Number MFCD00075635
InChI Key GKFJEDWZQZKYHV-UHFFFAOYSA-N
Synonym tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane
PubChem CID 6364547
IUPAC Name boron;2-methylpropan-2-amine
SMILES B.CC(C)(C)N

Specifications

Melting Point 96°C to 98°C
Density 0.87
Quantity 10 g
UN Number UN2811
Beilstein 4134905
Solubility Information Soluble in water (27g/L at 20°C) and methanol.
Formula Weight 86.98
Percent Purity 97%
Chemical Name or Material Borane-tert-butylamine complex

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.