missing translation for 'onlineSavingsMsg'
Learn More

Borane-tetrahydrofuran complex, 1M soln. in THF, stab. with 5mmol NaBH4, Thermo Scientific™

Product Code. 11486800
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486800 25 mL 25mL
15143726 100 mL 100mL
11406810 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486800 Supplier Thermo Scientific Alfa Aesar Supplier No. L13961.AC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Borane-tetrahydrofuran complex is an important reagent used in the reduction of certain functional groups viz. aldehyde, ketone, carboxylic acid, amide, oxime, imine and nitrile. It is also used as hydro borating agent. It acts as a borane source for oxazaborolidine catalyzed asymmetric reductions. It is utilized to reduce nylon surface amide groups to secondary amines.

Solubility
Miscible and reacts violently with water.

Notes
Air and moisture sensitive. Forms explosive peroxides in contact with air. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and alcohols. On hydrolysis, it forms hydrogen and boric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14044-65-6
Molecular Formula C4H11BO
Molecular Weight (g/mol) 85.94
MDL Number MFCD00012429
InChI Key RMCYTHFAWCWRFA-UHFFFAOYSA-N
Synonym borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf
PubChem CID 11062302
IUPAC Name boron;oxolane
SMILES B.C1CCOC1

Specifications

Density 0.878
Flash Point −21°C (−8°F)
Linear Formula BH3/(CH2)4O
Quantity 25 mL
UN Number UN3148
Beilstein 3668402
Sensitivity Air and moisture sensitive
Solubility Information Miscible and reacts violently with water.
Formula Weight 85.94
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Chemical Name or Material Borane-tetrahydrofuran complex, stabilized with 5mmol NaBH4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.