missing translation for 'onlineSavingsMsg'
Learn More

Borax, MP Biomedicals™

Product Code. 11461831 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
11461831 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 11461831

Brand: MP Biomedicals 02190308.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Typically used as a buffer component. Borax, MP Biomedicals is a salt of boric acid. It has a co-complexing ability with other agents in water to form complex ions with various substances. It is also commonly utilized in chromatography and electrophoresis

Borax is typically used as a buffer component and in the manufacturing of glazes and enamels, in fireproofing of fabrics and wood, and in antiseptic and detergents.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1303-96-4
Quantity 500 g
Molecular Formula Na2B4O7. 10H2O
Physical Form White Powder
Synonym Borax decahydrate, Sodium borate decahydrate, Sodium tetraborate, Sodium borate, Sodium biborate, Sodium pyroborate, Sodium tetraborate decahydrate
Solubility Information Anhydrous is slowly soluble in water; Decahydrate is soluble in water (1g/16mL at room temperature; 1g/0.6mL boiling water) or glycerol (1g/mL at room temperature). Insoluble in ethanol.
Formula Weight 381.4
Percent Purity ∼99%
compliance-icons
Product Identifier
  • Borax
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.