missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride, 99.5% (metals basis)

Product Code. 15432047
Change view
Click to view available options
Quantity:
1 Pc.
Packaging:
Bag
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15432047 1 Pc. Bag Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15432047 Supplier Thermo Scientific Alfa Aesar Supplier No. 044839.KG
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10043-11-5 | BN | 24.82 g/mol

Boron nitride is used as polymorph, cosmetics, paints, dental cements, and pencil leads. It is also used in xerographic process and laser printers as a charge leakage barrier layer of the photo drum, fillers, processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10043-11-5
Physical Form Hot Pressed Rod
Quantity 1 Pc.
Molecular Formula BN
MDL Number MFCD00011317
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
Solubility Information Insoluble in water.
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
SMILES B=N
IUPAC Name azanylidyneborane
Molecular Weight (g/mol) 24.82
PubChem CID 66227
ChEBI CHEBI:50883
Formula Weight 24.82
Percent Purity 99.5%
Packaging Bag
Chemical Name or Material Boron nitride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.