missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific™

Product Code. 15401887
Change view
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15401887 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15401887 Supplier Thermo Scientific Alfa Aesar Supplier No. 043255.KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10043-11-5
Physical Form Hot Pressed Rod
Quantity 1 Pc.
Molecular Formula BN
MDL Number MFCD00011317
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
Solubility Information Insoluble in water. Boron nitride is insoluble in the acids. Soluble in alkaline molten salts and nitrides,
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
SMILES B=N
IUPAC Name azanylidyneborane
Molecular Weight (g/mol) 24.82
PubChem CID 66227
ChEBI CHEBI:50883
Formula Weight 24.82
Percent Purity 99.5%
Packaging Bag
Chemical Name or Material Boron nitride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.