missing translation for 'onlineSavingsMsg'
Learn More

Boron oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific™

Product Code. 11376178
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376178 500 g 500g
11366178 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376178 Supplier Thermo Scientific Alfa Aesar Supplier No. 011158.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
PubChem CID 518682
ChEBI CHEBI:30163
IUPAC Name oxo(oxoboranyloxy)borane
SMILES O=BOB=O

Spezifikation

Boiling Point 1860°C
Melting Point ∼450°C
Color White
Physical Form Fused Lump
Quantity 500 g
Merck Index 14,1337
Formula Weight 69.62
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic
Density 2.46 g/mL (±)0.01
Assay (metals basis)
Chemical Name or Material Boron oxide, Fused Lump, Approx. 1.0 in. dia. x 0.75 in. thick
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.