Learn More
Boron oxide, Puratronic™, (H{2}O 200ppm), 99.999% (metals basis), Thermo Scientific Chemicals
Descrizione
In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP).Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. It has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants, heatproof glasswares and dopants in semiconductor materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
Specifica
Specifica
| Boiling Point | 1860°C |
| Melting Point | ∼450°C |
| Color | White |
| Quantity | 10 x 55 g |
| Merck Index | 14,1337 |
| Formula Weight | 69.62 |
| Percent Purity | 99.999% |
| Grade | Puratronic |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Vedi altri risultati |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.