missing translation for 'onlineSavingsMsg'
Learn More

Boron powder, 98% (metals basis), Thermo Scientific Chemicals

Product Code. p-4407286
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436315 10 g 10g
15446315 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15436315 Supplier Thermo Scientific Alfa Aesar Supplier No. 040036.09
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-42-8
Molecular Formula B
Molecular Weight (g/mol) 10.81
MDL Number MFCD00134034 MFCD00151272
InChI Key ZOXJGFHDIHLPTG-UHFFFAOYSA-N
Synonym boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
PubChem CID 5462311
ChEBI CHEBI:27560
IUPAC Name boron
SMILES [B]

Specifications

Color Brown
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 98% (Metals basis)
Odor Odorless
Appearance Brown powder
Chemical Name or Material Boron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.