missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 99% min

Product Code. 15431357
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. Quantity unitSize
15431357 50 g 50g
15441357 250 g 250g
2 options
This item is not returnable. View return policy

Product Code. 15431357

Brand: Thermo Scientific Alfa Aesar 035846.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

As a catalyst in the manufacturing of diborane, ultra pure boron | CAS: 10294-33-4 | BBr3 | 250.52 g/mol

As a catalyst in the manufacturing of diborane, ultra pure boronBoron tribromide is used as a reactant for luminescent polystyrene derivatives with sterically protected carbazolylborane moieties, high-quality boron-doped graphene via

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-33-4
Molecular Formula BBr3
Molecular Weight (g/mol) 250.52
MDL Number MFCD00011312
InChI Key ILAHWRKJUDSMFH-UHFFFAOYSA-N
Synonym boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
PubChem CID 25134
IUPAC Name tribromoborane
SMILES BrB(Br)Br

Specifications

Boiling Point 90°C
Melting Point -46°C
Color Colorless to Orange
Physical Form Liquid
Quantity 50 g
UN Number UN2692
Merck Index 14,1347
Solubility Information Miscible with ethanol and carbon tetrachloride.
Formula Weight 250.52
Percent Purity ≤99%
Sensitivity Moisture sensitive
Density 2.643 g/mL
Chemical Name or Material Boron tribromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.