missing translation for 'onlineSavingsMsg'
Learn More

Boron trichloride, 1M soln. in dichloromethane, stab.

Product Code. 15418195
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
15418195 500 mL 500mL
15408195 100 mL 100mL
2 options
This item is not returnable. View return policy

Product Code. 15418195

Brand: Thermo Scientific Alfa Aesar H32751.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-34-5 | BCl3 | 117.16 g/mol

Boron trichloride is used in semiconductor production and in the preparation of other boron compounds. It serves as a reagent in the preparation of various protease inhibitors. It is also used as a catalyst and soldering flux. It is involved in the improvement of aluminum castings by treating with boron trichloride vapors. It is utilized to produce electrical resistors. It is also used as a metal refining agent, especially for aluminum, magnesium, zinc and copper alloys. Further, it is used in the preparation of nanomaterials such as nanofibers and nanotubes. In addition to this, it is used in the field of high energy fuels and rocket propellants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron trichloride is used in semiconductor production and in the preparation of other boron compounds. It serves as a reagent in the preparation of various protease inhibitors. It is also used as a catalyst and soldering flux. It is involved in the improvement of aluminum castings by treating with boron trichloride vapors. It is utilized to produce electrical resistors. It is also used as a metal refining agent, especially for aluminum, magnesium, zinc and copper alloys. Further, it is used in the preparation of nanomaterials such as nanofibers and nanotubes. In addition to this, it is used in the field of high energy fuels and rocket propellants.

Solubility
Miscible with dichloromethane, ethanol, carbon tetrachloride, diethyl ether, dimethyl formamide, aromatic solvents, saturated and halogenated hydrocarbon.

Notes
Moisture sensitive. Incompatible with water, oxidizing agents, bases and metals. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-34-5
Molecular Formula BCl3
Molecular Weight (g/mol) 117.16
MDL Number MFCD00011313
InChI Key FAQYAMRNWDIXMY-UHFFFAOYSA-N
Synonym boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
PubChem CID 25135
IUPAC Name trichloroborane
SMILES ClB(Cl)Cl

Specifications

Quantity 500 mL
UN Number UN2927
Solubility Information Miscible with dichloromethane,ethanol,carbon tetrachloride,diethyl ether,dimethyl formamide,aromatic solvents,saturated and halogenated hydrocarbon.
Formula Weight 117.17
Concentration or Composition (by Analyte or Components) 1M soln.
Sensitivity Moisture sensitive; May form a precipitate
Density 1.326
Chemical Name or Material Boron trichloride, stabilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.