missing translation for 'onlineSavingsMsg'
Learn More

Boron trichloride, 1M soln. in hexanes, mixed isomers

Product Code. 15428215
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
15428215 500 mL 500mL
15418215 100 mL 100mL
2 options
This item is not returnable. View return policy

Product Code. 15428215

Brand: Thermo Scientific Alfa Aesar H32833.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-34-5 | BCl3 | 117.16 g/mol

A mild, selective combination reagent for the cleavage of primary alkyl aryl ethers. Boron trichloride is a selective demethylating agent. For preparing methyl esters of fatty acids and for transesterification of triglycerides. Mainly used as semiconductor silicon doping source or organic synthesis catalyst, also used for making high purity boron or organic boron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A mild, selective combination reagent for the cleavage of primary alkyl aryl ethers. Boron trichloride is a selective demethylating agent. For preparing methyl esters of fatty acids and for transesterification of triglycerides. Mainly used as semiconductor silicon doping source or organic synthesis catalyst, also used for making high purity boron or organic boron.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, heat. Unstable. Store at 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-34-5
Molecular Formula BCl3
Molecular Weight (g/mol) 117.16
MDL Number MFCD00011313
InChI Key FAQYAMRNWDIXMY-UHFFFAOYSA-N
Synonym boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
PubChem CID 25135
IUPAC Name trichloroborane
SMILES ClB(Cl)Cl

Specifications

Quantity 500 mL
Linear Formula BCl3
UN Number UN3286
Solubility Information Reacts with water.
Formula Weight 117.17
Concentration or Composition (by Analyte or Components) 1M soln. in hexanes
Sensitivity Moisture sensitive; May form a precipitate
Density 0.738
Chemical Name or Material Boron trichloride, mixed isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.