missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride dimethanol complex, 50-52 wt% BF3

Product Code. 10387570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10387570 250 g Glass bottle 250g
10091990 50 g Glass bottle 50g
10307530 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10387570 Supplier Thermo Scientific Acros Supplier No. 158902500
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2802-68-8 | C2H8BF3O2 | 131.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2802-68-8 , 7637-07-2 , 67-56-1
Molecular Formula C2H8BF3O2
Molecular Weight (g/mol) 131.89
MDL Number MFCD00071635
InChI Key MPRMFRXCVMCOMX-UHFFFAOYSA-N
Synonym boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
PubChem CID 11062313
SMILES CO.CO.FB(F)F

Specifications

Melting Point -20.0°C
Color Colorless to Yellow
Density 1.2220g/mL
Boiling Point 59.0°C (4.0 mmHg)
Flash Point 68°C
Packaging Glass bottle
Linear Formula BF3·2CH3OH
Refractive Index 1.34
Quantity 250 g
Specific Gravity 1.222
Solubility Information Solubility in water: hydrolysis.
IUPAC Name bis(methanol); trifluoroborane
Formula Weight 131.89
Percent Purity ≥50% (BF3)
Physical Form Liquid
Chemical Name or Material Boron trifluoride dimethanol complex, 50-52 wt% BF3
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Boron trifluoride dimethanol complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Methyl alcohol, Borate(1-), trifluoromethoxy-, (T-4)-, hydrogen, compound with methanol (1:1), Boron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.