missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride dimethyl etherate

Product Code. 10512421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10512421 50 mL Glass bottle 50mL
10368290 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10512421 Supplier Thermo Scientific Acros Supplier No. 204060500
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 353-42-4 | C2H6BF3O | 113.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-42-4
Molecular Formula C2H6BF3O
Molecular Weight (g/mol) 113.87
MDL Number MFCD00013192
InChI Key KTPWETRNSUKEME-UHFFFAOYSA-N
Synonym boron trifluoride dimethyl etherate, boron trifluoride methyl etherate, dimethyl ether trifluoroboron, dimethyloxonio trifluoro boranuide, boron trifluoride dimethyl ether complex, dimethyloxo trifluoro-??-borane
PubChem CID 2724165
IUPAC Name dimethyloxonio(trifluoro)boranuide
SMILES COC.FB(F)F

Specifications

Melting Point -14.0°C
Color Brown
Density 1.2300g/mL
Boiling Point 126.0°C to 127.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)2O·BF3
Refractive Index 1.3070 to 1.3090
Quantity 50 mL
Specific Gravity 1.23
Solubility Information Solubility in water: reacts
Formula Weight 113.87
Percent Purity ∼60% (BF3)
Physical Form Liquid, Fuming In Moist Air
Chemical Name or Material Boron trifluoride dimethyl etherate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Boron trifluoride dimethyl etherate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.