missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride etherate, ca. 48% BF3, AcroSeal™

Product Code. 10396814 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10396814 800 mL AcroSeal™ Glass Bottle 800mL
10304034 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396814 Supplier Thermo Scientific Acros Supplier No. 427118000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-63-7 | C4H10BF3O | 141.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 109-63-7
Molecular Formula C4H10BF3O
Molecular Weight (g/mol) 141.93
MDL Number MFCD00013194
InChI Key KZMGYPLQYOPHEL-UHFFFAOYSA-N
Synonym Boron trifluoride ethyl ether
PubChem CID 8000
IUPAC Name ethoxyethane;trifluoroborane
SMILES FB(F)F.CCOCC

Spezifikation

Melting Point -60.0°C
Color Colorless to Straw-Yellow
Density 1.1200g/mL
Boiling Point 126.0°C
Flash Point 58°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (C2H5)2O·BF3
Refractive Index 1.3429 to 1.3449
Quantity 800 mL
Merck Index 15, 1352
Specific Gravity 1.12
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol,soluble in ether
Viscosity 1.89 mPa.s (20°C)
Formula Weight 141.93
Percent Purity ca. 48% BF3
Physical Form Liquid
Chemical Name or Material Boron trifluoride etherate, ca. 48% BF3
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.