Learn More
Fisherbrand™ Borosilicate Glass Conical Shape Narrow Neck Erlenmeyer Flasks
Product Code. 12967620 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
50 mL
500 mL
3000 mL
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12967620 3000 mL Each
11557422 500 mL Pack of 10
11527422 50 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12967620
Quantity Price
1 48.35 € / Pack of 1
6 43.52 € / Pack of 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Erlenmeyer, graduated.
Specifications
Certifications/Compliance ISO 1773
Diameter (Metric) Outer Neck 22 mm
Capacity (Metric) 3000 mL
Material Borosilicate Glass
Shape Conical
Mouth Style Narrow