Learn More
Quickfit™ Borosilicate Glass Nitrogen (Kjeldahl) Determination Assembly
Product Code. 12094607
Change view
Click to view available options
Material:
Pyrex Borosilicate Glass
Unit Size:
Each
Product Code. Material Unit Size
12094607 Pyrex Borosilicate Glass Each
1 options
Product Code. 12094607 Supplier Quickfit™ Supplier No. 280MC
  • ONLINE EXCLUSIVE PRICE
    649.00 €
    534.38 € / Pack of 1
    Save 114.62 €
    17% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Complete assembly for nitrogen determination via the Kjeldahl method. Quickfit™ Borosilicate Glass Nitrogen (Kjeldahl) Determination Assembly is manufactured from heat-resistant and chemically durable borosilicate glass.
Complete assembly for nitrogen determination via the Kjeldahl method. Quickfit™ Borosilicate Glass Nitrogen (Kjeldahl) Determination Assembly is manufactured from heat-resistant and chemically durable borosilicate glass.
  • Convenient assembly of Quickfit™ glassware for Kjeldahl digestions and distillations
  • Flask can be used alone for digestion stage and then fitted to rest of assembly for the distillation process
  • Manufactured from heat resistant and chemically durable borosilicate glass
Specifications
Includes RA1/22 Adapter receiver, straight typeC1/12/SC Liebig condenserSH7/12 Adapter splash head, vertical distillationMA1/3 Adapter multiple with two parallel necksD3/12 Funnel dropping, open top with Rotaflo™ stopcockFK500/3L Kjeldahl flask 500ml
For Use With (Application) For Nitrogen determination
Type Kjeldahl distillation assembly
Material Pyrex Borosilicate Glass