missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ Borosilicate Glass Vials

Product Code. 12387059
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12387059

Brand: JG Finneran Associates™ 32008E1232

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Specifications

Color Clear
Material Borosilicate Glass
Volume (Metric) 2 mL
For Use With (Application) Designed to work in a variety of autosamplers requiring narrow neck vials.
Graduated Yes
No. per Pack 100/Pk.
Quantity 100/Pk.
Closure Size 8-425
Marking Spot White
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.