missing translation for 'onlineSavingsMsg'
Learn More

Bortezomib Impurity 11, TRC

Product Code. 30345021
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345021 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345021 Supplier Toronto Research Chemicals Supplier No. TRCB9739101MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bortezomib Impurity 11
CAS 862895-00-9
Molecular Formula C19H22N4O2
Quantity 1 mg
Synonym N-[(1S)-2-[[(1E)-3-Methyl-1-buten-1-yl]amino]-2-oxo-1-(phenylmethyl)ethyl]-2-pyrazinecarboxamide
InChI Formula InChI=1S/C19H22N4O2/c1-14(2)8-9-22-18(24)16(12-15-6-4-3-5-7-15)23-19(25)17-13-20-10-11-21-17/h3-11,13-14,16H,12H2,1-2H3,(H,22,24)(H,23,25)/b9-8+/t16-/m0/s1
SMILES O=C(NC(C(=O)NC=CC(C)C)CC=1C=CC=CC1)C2=NC=CN=C2
IUPAC Name N-[(2S)-1-[[(E)-3-methylbut-1-enyl]amino]-1-oxo-3-phenylpropan-2-yl]pyrazine-2-carboxamide
Molecular Weight (g/mol) 338.4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.