missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Bottle Top Dispensers

Product Code. 16892080 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
0.25 to 2.5 mL
0.5 to 5 mL
1 to 10 mL
10 to 100 mL
2.5 to 25 mL
5 to 50 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
16892080 0.25 to 2.5 mL Each
16802090 0.5 to 5 mL Each
16812090 1 to 10 mL Each
16822090 2.5 to 25 mL Each
16832090 5 to 50 mL Each
16842090 10 to 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16892080 Supplier Corning™ Supplier No. 6840
  • ONLINE EXCLUSIVE PRICE
    452.00 €
    421.88 € / Pack of 1
    Save 30.12 €
    6% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Corning™ Bottle Top Dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent.

  • Accurate and reproducible volume setting
  • 360° rotatable valve block for convenient dispensing with tough bottle connections
  • Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
  • Unique drip-free system to increase safety at work
  • Easy disassembly for cleaning
  • Easy in-lab calibration with built-in recalibration mechanism
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±0.012 mL
Product Type Bottletop Dispenser
Certifications/Compliance ISO 8655
Color Gray, Orange
Density 2.2 g/cm3
Material Polypropylene, PTFE
Volume (Metric) 0.25 to 2.5 mL
For Use With (Application) Quick and precise dispensing of acids, bases and solvents
For Use With (Equipment) Glass and Plastic Bottles
Includes Calibration Tool, Thread Adaptors Set (4 sizes), Telescopic Suction Tube, Instruction Manual, Certificate of Performance
Increments 0.05 mL
Precision ≤ 0.002 mL
Sterility Non-sterile
Temperature (English) 33.8°F to 104°F
Temperature (Metric) 1°C to 40°C
Vapor Pressure ≤ 500 mbar
Viscosity Rating ≤ 500 mm2/sec
Warranty 3 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.