missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine alpha-Thrombin Native Protein

Product Code. 11504141
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11504141 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11504141 Supplier Invitrogen™ Supplier No. RP43104
  • ONLINE EXCLUSIVE PRICE
    151.00 €
    134.39 € / 200µg
    Save 16.61 €
    11% Off
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Native Protein

RP-43104 contains native bovine a-Thrombin protein. RP-43104 may be used as a positive control in many immunological assays.

Note: Antibody lot-to-lot variability may occur and dilution optimization of each production lot in specific application is recommended.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00735
Concentration 9.2 mg/mL
For Use With (Application) Neutralization
Formulation 50% water with 50% glycerol and no preservative
Gene ID (Entrez) 280685
Molecular Weight (g/mol) 36.7 kDa
Name Bovine alpha-Thrombin
Purification Method SDS-PAGE
Quantity 200 μg
Source Bovine
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Activation peptide fragment 1; Activation peptide fragment 2; bovine prothrombin protein; Coagulation factor II; coagulation factor II (thrombin); coagulation factor II [thrombin]; coagulation factor II, thrombin; F2; prepro-coagulation factor II; preprothrombin; prothrombin; prothrombin B-chain; PT; RPRGL2; serine protease; THPH1; thrombin; Thrombin heavy chain; Thrombin light chain
Common Name alpha-Thrombin
Gene Symbol F2
Product Type Protein
Conjugate Unconjugated
Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.