missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine anti-Chicken IgY Secondary Antibody, Agarose

Product Code. 11505190
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11505190 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11505190 Supplier Invitrogen™ Supplier No. SA19588
  • ONLINE EXCLUSIVE PRICE
    516.00 €
    459.24 € / 1mL
    Save 56.76 €
    11% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bovine Polyclonal Secondary Antibody

SA1-9588 contains 1 mL of antibody linked to agarose beads (50%) in PBS with 0.02% sodium azide. This antibody reacts with the heavy and light chains of the immunoglobulin molecule. SA1-9588 immunogen is immunoglobulin isolated from chicken serum. To obtain optimal immunoprecipitation, incubate the beads with samples containing IgY antibodies-protein complex for at least 10 minutes. Binding capacity of the beads is approximately 450 μg to 500 μg IgY per mL solid beads.

Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chicken IgY
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Agarose
Formulation PBS with 0.02% sodium azide
Host Species Bovine
Immunogen Immunoglobulin isolated from chicken serum.
Purification Method Protein A/G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chicken
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Slurry
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.