missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine Factor IXa Native Protein

Product Code. 11574141
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11574141 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11574141 Supplier Invitrogen™ Supplier No. RP43112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Native Protein

RP-43112 contains native bovine Factor Ixa protein. RP-43112 may be used as a positive control in many immunological assays.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00741
For Use With (Application) Control
Formulation 50% water with 50% glycerol and no preservative
Gene ID (Entrez) 280688
Molecular Weight (g/mol) 45 kDa
Name Bovine Factor IXa
Quantity 100 μg
Source Bovine
Regulatory Status RUO
Gene Alias AW111646; Cf9; Cf-9; Christmas factor; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; F9; F9 p22; F9a; factor 9; factor IX (Christmas factor) precursor; factor IX F9; FIX; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; THPH8
Common Name Factor Ixa
Gene Symbol F9
Conjugate Unconjugated
Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.