missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine Factor Va Native Protein

Product Code. 11514161
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11514161 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11514161 Supplier Invitrogen™ Supplier No. RP43129
  • ONLINE EXCLUSIVE PRICE
    560.00 €
    498.40 € / 100µg
    Save 61.60 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Native Protein

RP-43129 contains native bovine Factor Va protein. RP-43129 may be used as a positive control in many immunological assays.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q28107
Concentration 2.4 mg/mL
For Use With (Application) Bioactivity
Formulation Protein with 5mM CaCl2, 50% glycerol and no preservative
Gene ID (Entrez) 280687
Molecular Weight (g/mol) 168 kDa
Name Bovine Factor Va
Purification Method SDS-PAGE
Quantity 100 μg
Source Bovine
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Activated protein C cofactor; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; F5a; factor V; factor V Leiden; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2
Common Name Factor Va
Gene Symbol F5
Biological Activity Specific Activity = 1792 U/mg; determined by Factor V clotting assay where 1 U equals the Factor V activity in 1 mL of normal human plasma.
Product Type Protein
Conjugate Unconjugated
Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Specific Activity = 1792 U/mg; determined by Factor V clotting assay where 1 U equals the Factor V activity in 1 mL of normal human plasma.
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.