Learn More
Edulab Boyles Law Apparatus
Product Code. 30280272
Change view
Click to view available options
Product Type:
Boyle's Law Demonstration Apparatus
Unit Size:
Each
Product Code. Product Type Unit Size
30280272 Boyle's Law Demonstration Apparatus Each
1 options
Product Code. 30280272 Supplier Edulab Supplier No. VTN12302225
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Demonstrates Boyle's Law in a clear and practical way for laboratory experiments.
  • Improved Boyle's Law apparatus designed for effective demonstration of pressure-volume relationships
  • Reservoir system allows colored water or oil to be pumped gradually into the tube containing air
  • Graduated tube enables accurate volume readings while pressure is measured using a reliable Bourdon gauge
Specifications
For Use With (Application) Demonstration of Boyle's Law
Includes One Apparatus
Product Type Boyle's Law Demonstration Apparatus