missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BPTES
GREENER_CHOICE

Product Code. 18787256
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18787256

Brand: Tocris Bioscience™ 5301/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glutaminase Inhibitors

BPTES is an allosteric glutaminase (GLS1) inhibitor (IC50 = 3.3 μM). Selective for GLS1 over GLS2 and γ-glutamyl transpeptidase. Induces cell death of P493 human lymphoma B cells in vitro and reduces tumor volume of P493 cell xenografts in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N′-[Thiobis(2,1-ethanediyl-1,3,4-thiadiazole-5,2-diyl)]bisbenzeneacetamide
CAS 314045-39-1
Quantity 10 mg
Target Hydrolase Inhibitors
Molecular Formula C24H24N6O2S3
Formula Weight 524.68
Purity >95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.