missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BPTU
GREENER_CHOICE

Product Code. 18768629
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18768629 10 mg 10mg
18723008 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18768629 Supplier Tocris Bioscience™ Supplier No. 6078/10
  • ONLINE EXCLUSIVE PRICE
    329.00 €
    310.80 € / 10mg
    Save 18.20 €
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

P2Y1 allosteric antagonist

BPTU is an allosteric antagonist of P2Y1 (EC50 = 0.06-0.3 μM). Non-nucleotide ligand. Binds receptor outside of the helical bundle. Blocks inhibition of spontaneous contraction of rat and mouse colon induced by electrical field stimulation, nicotine and P2Y agonists. Antithrombotic; reduces platelet aggregation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-[2-(1,1-Dimethylethyl)phenoxy]-3-pyridinyl]-N′-[4-(trifluoromethoxy)phenyl]urea
CAS 870544-59-5
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C23H22F3N3O3
Formula Weight Observed MW: 445.43
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.