missing translation for 'onlineSavingsMsg'
Learn More

BQ-123, MedChemExpress

Product Code. 16477847
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477847 5 mg 5mg
16455128 10 mg 10mg
16401249 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16477847 Supplier Medchem Express Supplier No. HY12378

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BQ-123 is a potent and selective endothelin A (ETA) receptor antagonist with an IC50 of 7.3 nM and a Ki of 25 nM. BQ-123 inhibits endothelin-1-mediated proliferation of human pulmonary artery smooth muscle cells and lowers blood pressure in different rat models of hypertension.

BQ-123 is a potent and selective endothelin A (ETA) receptor antagonist with an IC50 of 7.3 nM and a Ki of 25 nM. BQ-123 inhibits endothelin-1-mediated proliferation of human pulmonary artery smooth muscle cells and lowers blood pressure in different rat models of hypertension.

Specifications

Chemical Name or Material BQ-123
CAS 136553-81-6
Color White
Purity Grade Notes Research
Molecular Formula C31H42N6O7
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (81.87 mM; Need ultrasonic)
SMILES O=C(N[C@](C(N(CCC1)[C@]1([H])C2=O)=O)([H])CC(O)=O)[C@H](NC([C@@H](NC([C@H](N2)C(C)C)=O)CC(C)C)=O)CC3=CNC4=CC=CC=C34
Molecular Weight (g/mol) 610.7
Formula Weight 610.7
Percent Purity 95.3%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.