Learn More
Brady™ BradyBondz™ WorkHorse™ Series Thermal Transfer Printable Label
BradyBondz™ WorkHorse™ Thermal Transfer Printable Clear Polyester (B-430) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos and require nameplate-like quality.
Brand:  Brady™ 030189
Product Code. 13581460
  • ONLINE EXCLUSIVE PRICE
    377.00 €
    360.03 € / 1 rolls
    Save 16.97 €
    4% Off
In Stock
Add to basket
Description
Description
  • Glossy, clear labels are thermal transfer printable and feature a durable permanent acrylic adhesive
  • Excellent resistance to most chemicals, oils and water
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Service temperature of -70 to 100° F (-56 to 137° C)
Specifications
Specifications
Autoclavable
Monochrome
Permanent
Gloss
Polyester
Non-sterile
Brady 1244, Brady 1344, Brady 2461, Brady 3481, Brady 6441, Brady 300MVP Plus, Brady 200MVP Plus, Brady 300X-Plus II, Brady 360X-Plus II, Brady 600X-Plus II, Tagus T200, Tagus T300, Thermal Transfer Printer, IP300 Thermal Transfer Printer, IP600 Thermal Transfer Printer, BBP81 Label Printer
3000/Roll
-70°C to +100°C
0.076 mm
3 in.
1 in.
3 in.
THT-18-430-3
UL Recognized, CSA Approved
Thermal Transfer Printable Labels
Clear
Roll
Thermal Transfer
Asset tracking, Barcoding, Component ID, Equipment ID, General purpose ID, Rating plates
THT-18-430-3
Rectangle
0.003 in.
1 x 3 in.
76.2 mm
25.4 mm
76.20 mm