missing translation for 'onlineSavingsMsg'
Learn More

Abcam Bradford Reagent, Protein Quantification, Liquid, 500 mL

Product Code. 32118940 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32118940 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32118940 Supplier Abcam Supplier No. ab119216500mL
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Improved protein quantification reagent with higher detergent tolerance and reduced protein-to-protein variation.

Bradford reagent is an improved formulation for protein quantification with higher tolerance to detergent contamination and reduced protein-to-protein variation.

  • Form: Liquid
  • Colorimetric detection: Binds to protein, causing a shift in absorption from 465 nm to 595 nm (color change from brown to blue)
  • Performance: Excellent linearity across a defined range of protein concentrations
  • Storage buffer constituents: 14% Phosphoric acid, 3% Ethanol, 2.3% α-Cyclodextrin hydrate, 0.23% gamma-Cyclodextrin, 0.005% Brilliant Blue G
  • Usage: Cited in over 30 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 14% phosphoric acid, 3% ethanol, 2.3% α-cyclodextrin hydrate, 0.23% gamma-cyclodextrin, 0.005% Brilliant Blue G
Content And Storage Store at 4°C.
Detection Location In-Solution Detection
Format Bottle
Form Liquid
Kit Contents Bradford Reagent (500 mL)
Shipping Condition Ambient
Substrate Protein
Target Molecule Protein
Detection Method Colorimetric
Formulation Constituents: 14% Phosphoric acid, 3% Ethanol, 2.3% α-Cyclodextrin hydrate, 0.23% gamma-Cyclodextrin, 0.005% Brilliant Blue G
Quantity 500 mL
For Use With (Application) Protein quantification
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.