missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Accessories for BMP71 Label Printer

Product Code. 11554094 Shop All Brady Products
Change view
Click to view available options
Product Type:
Hard Case
Rechargeable Battery Pack
Soft Case
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11554094 Rechargeable Battery Pack Each
12880009 Soft Case Each
11899514 Hard Case Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11554094 Supplier Brady™ Supplier No. 114885
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Printer case, Quick charger and Printer battery for use with BMP-71 Label Printer (printer sold separately).

  • BMP™71 Label Printer Accessories from Brady™
  • Printer Case designed to protect your printer against impacts and dust while allowing for easy mobility.
  • Printer Battery offers long battery life - prints up to 3900 1 in. x 2 in. labels per charge, enables cordless printer operation so you can print labels where you need them, when you need them.
  • BMP™71 Quick Charger, charges battery in one hour.

Specifications

Product Type Rechargeable Battery Pack
Color Gray
For Use With (Equipment) BMP71 Label Printer
Sterility Non-sterile
Description Spare Battery
Quantity 1 Ea.
Alternate Part Number M71-BATT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.