missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Battery Pack for Brady Mobile Printers

Product Code. 11952742 Shop All Brady Products
Change view
Click to view available options
Color:
Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11952742 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11952742 Supplier Brady™ Supplier No. 142260
  • ONLINE EXCLUSIVE PRICE
    230.00 €
    220.71 € / Pack of 1
    Save 9.29 €
    4% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Battery Pack for use with Brady™ BMP51 and BMP53 printers.

  • Package includes spare Li-Ion Printer Battery for the BMP51 and BMP53 printers only (label printer and AC Adaptor sold separately)
  • Long battery life - prints up to 1000 1 in. x 2 in. labels per battery charge
  • Use AC adapter/charger unit (not included) to charge battery
  • Enables cordless printer operation so you can print labels where you need them, when you need them

Specifications

Product Type Rechargeable Li-ION Battery
Color Gray
For Use With (Equipment) Brady Mobile Printers
Sterility Non-sterile
Alternate Part Number BMP-UBP 12V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.