Learn More
Brady™ BBP™ Repositional Vinyl Film
BBP™ Repositional Vinyl Film (B-581) is a durable, flexible, high gloss material ideal for indoor marking where frequent repositioning is required.
Brand:  Brady™ 118009
 View more versions of this product
Product Code. 11766465
  • ONLINE EXCLUSIVE PRICE
    190.00 €
    181.68 € / Pack of 1
    Save 8.32 €
    4% Off
Estimated Shipment: 08-10-2026
Add to basket
Description
Description
  • Special aggressive adhesive stays in place but removes cleanly without leaving adhesive residue making it ideal for applications like 5S and inventory
  • Vinyl film can be thermal transfer printed
  • Average outdoor durability is 5 years
  • Service temperature of -40°F to 180°F (-40°C to 82°C)
Specifications
Specifications
Acrylic, Repositionable, Pressure-sensitive
Gloss
Vinyl
Ideal for temporary, Removable and repositionable labels, like 5S and lean inventory label
5 Years
-40 to 180°F
0.005 in.
30.48 m
B30C-1125-581-YL
Yellow
Film
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1 Roll
-40°C to +82°C
100 ft.
1.12 in.