Learn More
Brady™ BBP™ Retro Reflective Tape
Highly reflective identification that shines back brightly when struck by light
Brand:  Brady™ 117997
 View more versions of this product
Product Code. 11746455
  • ONLINE EXCLUSIVE PRICE
    434.00 €
    417.27 € / Pack of 1
    Save 16.73 €
    3% Off
Estimated Shipment: 13-10-2026
Add to basket
Description
Description
  • Ideal for labeling outdoor utility poles and pedestals, jobsite equipment and roof-top equipment
  • Recommended low light or inclement weather conditions
  • Available in multiple colors
Specifications
Specifications
Retro Reflective Tape for BBP3x Printers
Silver
Tape
For labeling outdoor utility poles and pedestals, jobsite equipment, roof-top equipment
100 Gardner Units
4 to 6 Years
Brady Workstation, LabelMark, MarkWare
-40°C to +70°C
0.0055 in.
50 ft.
4 in.
B30C-4000-584-SL
Acrylic, Pressure-sensitive
Gloss, Reflective
Polyester
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1
1 Roll
-40 to 158°F
20 lb./in.
0.14 mm
15.24 m
101.60 mm