missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Hazwik™ Spill Kits

Product Code. 10409894
Change view
Click to view available options
Capacity (English):
5 gal.
Unit Size:
Each
Product Code. Capacity (English) unitSize
10409894 5 gal. Each
1 options
This item is not returnable. View return policy

Product Code. 10409894

Brand: Brady™ 813858

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical class spill kits

SKH-CFB Emergency Response Portable Spill Kit

  • Reusable clear bag with shoulder strap
  • (32) 38.1 x 48.26cm (15 x 19 in.) pads
  • (5) 7.62cm x 1.22m (3 in. x 4 ft.) SOCs
  • 43.18 x 48.26cm (17 x 19 in.) pillow
  • Nitrile gloves, goggles, two disposal bags and instruction sheet
  • Shipping wt: 11.34kg (25 lbs.)

SKH-PP — Portable Economy Spill Kit

  • For small spills of hazardous materials
  • (10) 38.1 x 48.26cm (15 x 19 in.) pads
  • (2) 7.62cm x 1.22m (3 in. x 4 ft.) SOCs
  • Nitrile gloves, disposal bag and instruction sheet
  • Shipping wt: 2.26kg (5 lbs.)

Specifications

Color Yellow
Capacity (English) 5 gal.
Capacity (Metric) 15.12 L
Material PVC
Type Portable Spill Containment Kit
For Use With (Application) Portable economy kit for small spills
Includes (10) 38.1 x 48.26 cm pads, (2) 7.62 cm x 1.22 m socks, nitrile gloves, disposal bag, instruction sheet
Packaging Quantity 1 Kit
Dimensions (L x W x H) 20 x 16 x 4 in. (50.8 x 40.64 x 10.16 cm)
Length (English) 20 in.
Length (Metric) 50.8 cm
Width (English) 16 in.
Width (Metric) 40.64 cm
Height (English) 4 in.
Height (Metric) 10.16 cm
Weight (English) 3.909 lb.
Weight (Metric) 1.8 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.