Learn More
Brady™ Labels for Desktop Laser Printers
Labels for Desktop Laser Printers ideal for rating plates, asset identification, and general purpose labeling applications.
Brand:  Brady™ 089405
 View more versions of this product
Product Code. 10407272
  • ONLINE EXCLUSIVE PRICE
    110.00 €
    104.53 € / Pack of 1
    Save 5.47 €
    4% Off
Estimated Shipment: 13-01-2027
Add to basket
Description
Description
  • Durable, chemical- and solvent-resistant labels for use with a variety of laser printers.
  • High resolution, good solvent and smudge resistance, moderate to high temperature performance.
  • Withstand extreme laboratory environments.
Specifications
Specifications
Autoclavable
LaserTab Laser Printable Labels
White
Matte
Nylon Cloth
Laser
Non-sterile
Laser Printers
Pack of 9 Sheets
-40°C
2500 Pk.
0.375 in. dia.
-40°C to +90°C
0.165 mm
0.375 in.
9.52 mm
0.37 in.
LAT-60-799-2.5
Halogen Free (DIN VDE 0472/part 815)
Acrylic, Permanent
Consignment
Label
Single-side Printable
Label
Freezers, Liquid Nitrogen, Autoclaves, Hot Water Baths, Laboratory Labeling and Identification
12.70 mm
15
2565 Labels/Pack
Round
-40°F to +194°F
6.5 mil
0.375 x 0.375 in. (9.35 x 9.35 mm)
0.375 in.
12.70 mm
0.952 cm