missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Signs: Keep Aisles Clear

Product Code. 11716365
Change view
Click to view available options
Color:
Blue on White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11716365 Blue on White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11716365 Supplier Brady™ Supplier No. 104508
  • ONLINE EXCLUSIVE PRICE
    138.00 €
    132.26 € / Pack of 1
    Save 5.74 €
    4% Off
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

KEEP AISLES CLEAR ToughStripe™ Floor Signs feature a highly visible, glossy appearance for communicating important safety messages and anti-skid sections to help prevent slips, trips and falls.

  • ToughStripe™ Floor Signs communicate important safety and warning messages to your employees in areas with foot traffic
  • Durable ToughStripe™ Anti-Skid Polyester (B-534) includes a rigid, clear overlaminate and is selectively covered by anti-skid tread
  • Highly visible, glossy material resists harsh chemicals and is easy to apply and remove
  • Floor signs alert workers of potential hazards, direct traffic, and enforce protective equipment requirements.

Specifications

Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Alternate Part Number B534FS 17 in. DIA BK,BL/WT KEEP AISLES CLEAR
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Rubber-based
Description ToughStripe - Pre-printed Floor Signs
Diameter (English) 17 in.
Diameter (Metric) 431.80 mm
For Use With (Application) Used to mark aisles, passageways, storage locations, point out safety equipment and exits and enforce protective equipment requirements
Max. Temperature (English) 129.9°F
Min. Temperature (English) -0.4°F
Thickness 0.01 in.
Temperature Range (Metric) -18°C to 54°C
Temperature Range (English) -0.4°F to 129.9 °F
Languages English
Displayed Text Keep Aisles Clear
Product Type Floor Safety Sign
Color Blue on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.