missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BradyJet™ J2000 Polypropylene Labels

Product Code. 17170391
Click to view available options
Size (English):
1 x 3 in.
100 ft. x 1.125 in.
3.35 x 2.17 in.
4.33 x 2.95 in.
6.1 x 4.13 in.
Unit Size:
1 roll
Each
Pack of 1000
Pack of 200
Pack of 270
This item is not returnable. View return policy

Product Code. 17170391

Brand: Brady™ 150545

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical resistant, low temperature labels ideal for full-color identification and barcode applications

  • This B-7425J polypropylene material is ideal for full-color identification and barcode applications where resistance to liquid nitrogen, autoclave conditions and harsh chemicals is required
  • Matte white polypropylene with acrylic adhesive is inkjet printable
  • Perfect for laboratory identification where labels may come in contact with harsh chemicals
  • Service temperature of -112 to 158° F (-80 to 70° C)
  • Specifications

    Autoclavable Autoclavable
    Adhesive Type Acrylic, Permanent
    Coating None
    Color White
    Finish Matte
    Format Die-cut
    Material Polypropylene
    Printable Sides Single-side Printable
    Print Technology Ink Jet
    Product Type Label
    Sterility Non-sterile
    For Use With (Application) Barcode labelling, Cold temperature labelling, Component and equipment labelling, Hazard communication identification, Laboratory labelling
    For Use With (Equipment) BradyJet™ J2000
    Labels per Row 1
    Min. Operating Temperature (Metric) -80°C
    No. per Pack 1000 Labels/Bag
    Quantity 1000/Pk.
    Shape Rectangle
    Size (English) 1 x 3 in.
    Temperature Range (English) -112°F to +158°F
    Temperature Range (Metric) -80°C to +70°C
    Thickness (English) 0.005 in.
    Thickness (Metric) 0.127 mm
    Dimensions (LxW) 25.40 x 76.2 mm
    Length (English) 1 in.
    Width (English) 3.2 in.
    Width (Metric) 76.20 mm
    Height (English) 1 in.
    Height (Metric) 25.40 mm
    Alternate Part Number J20-18-7425J
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.