missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Bradylink™ Warning Post and Chain

Product Code. 11721955
Change view
Click to view available options
Color:
Black
Yellow
Quantity:
1 Ea.
6 Pk.
Unit Size:
Each
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
11721955 Black 1 Ea. Each
11741356 Black 6 Pk. Pack of 6
11761356 Yellow 6 Pk. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11721955 Supplier Brady™ Supplier No. 092116
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

This Bradylink™ Warning Post and Chain is made of colorful polystyrene and polyethylene and are a decorative alternative to metal posts.

  • Posts are non-conductive so can be safely used in electrically hazardous areas.
  • Unlike metal posts, Bradylink™ Warning Posts are non-rusting.
  • Hollow base can be filled with sand for more stable application.
  • Large C-hook holds chain in place on large posts and can be used open or closed for greater security.
  • Bases and tops are included.

Specifications

Product Type Warning Post and Chain
Alternate Part Number BRADYLINK WARNING POST SOLID BLACK -1 EA
Color Black
Material Polyethylene
Description Bradylink Warning Posts
Diameter (English) 3 in.
For Use With (Application) Traffic Control
Height (English) 41 in.
Height (Metric) 1041.4 mm
Includes Bases and tops
Quantity 1 Ea.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.