missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies BrainPhys™ hPSC Neuron Kit

Product Code. 17128301
Encompass
Change view
Click to view available options
Quantity:
500 mL kit
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17128301 500 mL kit 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17128301 Supplier STEMCELL Technologies Supplier No. 05795
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kit including BrainPhys™ Neuronal Medium, SM1 Neuronal Supplement, N2 Supplement-A, BDNF and GDNF for culture of ES/iPS cell-derived neurons

  • Promote, rather than inhibit, neuronal activity and maturity in your cultured primary or human pluripotent stem cell (hPSC)-derived neurons
  • Based on the formulation by Bardy and Gage (Bardy et al. PNAS, 2015), BrainPhys serum-free neuronal basal medium is optimized to yield a higher proportion of synaptically active neurons by mimicking the central nervous system (CNS) extracellular environment
  • Use BrainPhys Neuronal Medium for long-term culture of hPSC- and CNS-derived neurons and to perform functional assays, such as microelectrode array-based recordings or live-fluorescent imaging, without shocking your cells by changing media
  • To ensure cell survival in long-term culture, BrainPhys must be combined with an appropriate supplement
  • For your convenience, BrainPhys Neuronal Medium N2-A & SM1 Kit is available for hPSC-derived neurons and can be combined with lineage-specific growth factors and/or small molecules

Specifications

Cell Type Neural Cells, PSC-Derived, Neurons, Pluripotent Stem Cells
For Use With (Application) Cell Culture; Differentiation; Maintenance
Quantity 500 mL kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.